Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

87719

Supelco

Tetramethylammonium chloride

for electrochemical analysis, ≥99.0%

Synonym(s):

TMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(Cl)
CAS Number:
Molecular Weight:
109.60
Beilstein/REAXYS Number:
3911201
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
Product 87719 is not currently sold in your country. Contact Technical Service

assay

≥99.0% (AT)
≥99.0%

form

crystals

mp

>300 °C (lit.)

solubility

methanol: 0.1 g/mL, clear, colorless

SMILES string

[Cl-].C[N+](C)(C)C

InChI

1S/C4H12N.ClH/c1-5(2,3)4;/h1-4H3;1H/q+1;/p-1

InChI key

OKIZCWYLBDKLSU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Chronic 2 - Skin Irrit. 2 - STOT SE 1 Oral

target_organs

Central nervous system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 1

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Welcker et al.
Genes & development, 27(23), 2531-2536 (2013-12-04)
The Fbw7 tumor suppressor targets a broad network of proteins for ubiquitylation. Here we show critical functions for Fbw7 dimerization in regulating the specificity and robustness of degradation. Dimerization enables Fbw7 to target substrates through concerted binding to two suboptimal
Wei Shen et al.
Chemical communications (Cambridge, England), 49(73), 8114-8116 (2013-08-08)
A simple and low-cost colorimetric assay utilizing ferrofluidic nanoparticulate probes (FNPs) and a ligase for single-nucleotide polymorphism genotyping is described. Excellent sensitivity and selectivity were accomplished through the engagement of the FNPs and a ligase chain reaction.
Marco I Ries et al.
The Journal of biological chemistry, 288(52), 37289-37295 (2013-11-15)
Metabolic profiling and structural elucidation of novel secondary metabolites obtained from derived deletion strains of the filamentous fungus Penicillium chrysogenum were used to reassign various previously ascribed synthetase genes of the roquefortine/meleagrin pathway to their corresponding products. Next to the
Jacob O Fierer et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(13), E1176-E1181 (2014-03-19)
Individual proteins can now often be modified with atomic precision, but there are still major obstacles to connecting proteins into larger assemblies. To direct protein assembly, ideally, peptide tags would be used, providing the minimal perturbation to protein function. However
Chi Nguyen et al.
Nature, 505(7483), 427-431 (2013-12-24)
Acyl carrier protein (ACP) transports the growing fatty acid chain between enzymatic domains of fatty acid synthase (FAS) during biosynthesis. Because FAS enzymes operate on ACP-bound acyl groups, ACP must stabilize and transport the growing lipid chain. ACPs have a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service