Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

87267

Supelco

Tetraethylammonium bistrifluoromethanesulfonimidate

≥99.0%

Synonym(s):

Bistrifluoromethanesulfonimide tetraethylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)4N[N(SO2CF3)2
CAS Number:
Molecular Weight:
410.40
Beilstein/REAXYS Number:
8243917
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

assay

≥99.0% (T)
≥99.0%

mp

102-108 °C

SMILES string

CC[N+](CC)(CC)CC.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C8H20N.C2F6NO4S2/c1-5-9(6-2,7-3)8-4;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h5-8H2,1-4H3;/q+1;-1

InChI key

PBVQLVFWBBDZNU-UHFFFAOYSA-N

General description

Tetraethylammonium bistrifluoromethanesulfonimidate is a quaternary ammonium salt mainly used as an electrolyte.
Visit our Sensor Applications portal to learn more.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service