Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

86927

Supelco

Tetrabutylphosphonium hexafluorophosphate

≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4P(PF6)
CAS Number:
Molecular Weight:
404.40
Beilstein/REAXYS Number:
5895425
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

assay

≥99.0% (gravimetric)
≥99.0%

form

powder

mp

226-228 °C

solubility

methanol: 0.1 g/mL, clear, colorless

SMILES string

F[P-](F)(F)(F)(F)F.CCCC[P+](CCCC)(CCCC)CCCC

InChI

1S/C16H36P.F6P/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-7(2,3,4,5)6/h5-16H2,1-4H3;/q+1;-1

InChI key

QLQAFNAPINCKBM-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Novel organic electrolyte with high conductivity in high energy density electric double-layer capacitors (EDLC)[1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Ue et al.
Journal of the Electrochemical Society, 141, 2989-2989 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service