Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

86867

Sigma-Aldrich

Tetrabutylammonium phosphate monobasic

purum, ≥97.0% (T)

Synonym(s):

Tetrabutylammonium dihydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N[OP(OH)2O]
CAS Number:
Molecular Weight:
339.45
Beilstein/REAXYS Number:
5196532
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

purum

assay

≥97.0% (T)

form

solid

mp

151-154 °C (lit.)

SMILES string

OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.H3O4P/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;(H3,1,2,3,4)/q+1;/p-1

InChI key

ARRNBPCNZJXHRJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Mild acid[1]; Preparation of allylic phosphates[2]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A.I. Meyers et al.
Tetrahedron Letters, 26, 2047-2047 (1985)
M. Julia et al.
Tetrahedron, 42, 3841-3841 (1986)
Pieter van Dillewijn et al.
Applied and environmental microbiology, 74(21), 6820-6823 (2008-09-16)
Homogenous preparations of XenB of Pseudomonas putida, pentaerythritol tetranitrate reductase of Enterobacter cloacae, and N-ethylmaleimide reductase of Escherichia coli, all type II hydride transferases of the Old Yellow Enzyme family of flavoproteins, are shown to reduce the polynitroaromatic compound 2,4,6-trinitrotoluene
Chao Zhong et al.
Carbohydrate polymers, 94(1), 38-45 (2013-04-03)
In this article, a novel and high efficient solvent, tetra-n-Butylammonium Hydroxide (TBAH), was used for dissolution and isolation of straw cellulose from wheat straw. The composition analysis with gas chromatography (GC) and the spectroscopic characterization analysis conducted by X-Ray diffraction
Zhenfeng He et al.
Dalton transactions (Cambridge, England : 2003), 41(33), 10043-10051 (2012-05-29)
A new kind of organic-inorganic hybrid complexes based on polyoxometalate were synthesized through symmetrically grafting two adeninyl groups onto Anderson-type MnMo(6) clusters and encapsulating the clusters by organic surfactants. The resultant complexes exhibited thermal-induced dynamic self-assembly behaviors which greatly depended

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service