Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

86317

Supelco

Amberlyst® 16 wet

hydrogen form, strongly acidic

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12181503
NACRES:
SB.52

Quality Level

form

beads

crosslinking

12 % cross-linked

technique(s)

LPLC: suitable

loss

~50% loss on drying, 110°C

matrix

styrene divinylbenzene (macroporous)

matrix active group

sulfonic acid functional group

particle size

~700 μm

capacity

4.8 meq/g

separation technique

cation exchange

Application

especially for heterogeneous catalysis, with more resistance to thermal degradation

Legal Information

Amberlyst is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service