Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

85435

Supelco

Silylating mixture II according to Horning

for GC derivatization

Synonym(s):

N,O-Bis(trimethylsilyl)acetamide – chlorotrimethylsilane – 1-(trimethylsilyl)imidazole mixture, 1-(Trimethylsilyl)imidazole/BSA/TMCS 3/3/2 (v/v/v), N,O-Bis(trimethylsilyl)acetamide/TMSIM/TMCS 3/3/2 (v/v/v)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

grade

derivatization grade ((GC derivatization))
for GC derivatization

form

liquid

quality

ampule

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

Storage temp.

2-8°C

InChI

1S/C8H21NOSi2.C6H12N2Si.C3H9ClSi/c1-8(9-11(2,3)4)10-12(5,6)7;1-9(2,3)8-5-4-7-6-8;1-5(2,3)4/h1-7H3;4-6H,1-3H3;1-3H3

Inchi Key

QSBHIAMPMUWWFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

39.2 °F - closed cup

flash_point_c

4.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service