Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

85418

Supelco

Siloprene Crosslinking Agent K-11

Selectophore

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700

product line

Selectophore

form

liquid

storage temp.

2-8°C

General description

Visit our Sensor Applications portal to learn more.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service