Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

85378

Supelco

GC Stationary Phase

phase DC-550, bottle of 50 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

description

a methylphenylsilicon

form

liquid

packaging

bottle of 50 mL

parameter

20-350 °C

technique(s)

gas chromatography (GC): suitable

matrix active group

DC-550 phase

viscosity

~180 mPa.s, neat(20 °C)

General description

Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service