Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

85377

Supelco

GC Stationary Phase

phase DC-200, bottle of 250 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710

description

a methylsilicon

form

liquid

packaging

bottle of 250 mL

parameter

20-200 °C

technique(s)

gas chromatography (GC): suitable

matrix active group

DC-200 phase

General description

Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service