Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

84928

Sigma-Aldrich

Tris[4-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)phenyl]phosphine

>97.0% (GC)

Synonym(s):

Tris[4-(1H,1H,2H,2H-perfluorodecyl)phenyl]phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H24F51P
CAS Number:
Molecular Weight:
1600.60
Beilstein/REAXYS Number:
8824708
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

>97.0% (GC)

form

powder

mp

95-98 °C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCc1ccc(cc1)P(c2ccc(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)cc2)c3ccc(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)cc3

InChI

1S/C48H24F51P/c49-25(50,28(55,56)31(61,62)34(67,68)37(73,74)40(79,80)43(85,86)46(91,92)93)16-13-19-1-7-22(8-2-19)100(23-9-3-20(4-10-23)14-17-26(51,52)29(57,58)32(63,64)35(69,70)38(75,76)41(81,82)44(87,88)47(94,95)96)24-11-5-21(6-12-24)15-18-27(53,54)30(59,60)33(65,66)36(71,72)39(77,78)42(83,84)45(89,90)48(97,98)99/h1-12H,13-18H2

InChI key

MTUBRWIMDWTZST-UHFFFAOYSA-N

Other Notes

contains up to 15 mol% phospine oxide (31P-NMR). Phosphine ligand for preparing the Bannwarth catalyst (catalyst for Stille and Suzuki coupling)

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

target_organs

Liver

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helvetica Chimica Acta, 84, 735-735 (2001)
S. Schneider et al.
Angewandte Chemie (International Edition in English), 112, 4293-4293 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service