Skip to Content
MilliporeSigma

84910

Selenourea

purum, ≥97.0% (Se)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
NH2CSeNH2
CAS Number:
Molecular Weight:
123.02
EC Number:
211-129-9
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
1734744
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥97.0% (Se)

mp

210-215 °C (dec.)

storage temp.

2-8°C

SMILES string

NC(N)=[Se]

InChI

1S/CH4N2Se/c2-1(3)4/h(H4,2,3,4)

InChI key

IYKVLICPFCEZOF-UHFFFAOYSA-N



signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



B Mishra et al.
The journal of physical chemistry. A, 110(5), 1894-1900 (2006-02-03)
Pulse radiolysis coupled with absorption detection has been employed to study one-electron oxidation of selenomethionine (SeM), selenocystine (SeCys), methyl selenocysteine (MeSeCys), and selenourea (SeU) in aqueous solutions. Hydroxyl radicals (*OH) in the pH range from 1 to 7 and specific
K P Kalyanikutty et al.
Journal of nanoscience and nanotechnology, 7(6), 1916-1922 (2007-07-28)
Two-dimensional nanostructures in the form of ultra-thin crystalline films of CdSe and CuSe have been prepared at the organic-aqueous interface by reacting toluene solutions of metal cupferronates with an aqueous solution of N,N-dimethyl selenourea. The films have been examined using
Cristina Trujillo et al.
The journal of physical chemistry. B, 112(17), 5479-5486 (2008-04-09)
The gas-phase reactions between Ca(2+) and selenourea were investigated by means of electrospray/tandem mass spectrometry techniques. The MS/MS spectra of [Ca(selenourea)](2+) complexes show intense peaks at m/z 43, 121, 124, and 146 and assigned to monocations produced in different coulomb