Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

84886

Millipore

Selective Agar for pathogenic fungi

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171609
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 12.5 g/L
chloramphenicol, 0.05 g/L
cycloheximide, 0.4 g/L
D(+)-glucose, 10.0 g/L
soya flour peptone, 10.0 g/L

final pH

6.9±0.2 (25 °C)

application(s)

agriculture
clinical testing
environmental
food and beverages

microbiology

suitability

selective for Candida spp.
selective for molds (General Media)
selective for yeasts (General Media)

Application

For the isolation of pathogenic fungi, in particular dermatophytes, from heavily contaminated material.

Other Notes

Use of cycloheximide in the selective isolation of fungi pathogenic to man[1]

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Muta. 2 - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of cycloheximide in the selective isolation of fungi pathogenic to man.
L K GEORG et al.
The Journal of laboratory and clinical medicine, 44(3), 422-428 (1954-09-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service