Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

84539

Sigma-Aldrich

Sarcosine ethyl ester hydrochloride

≥99.0% (AT)

Synonym(s):

Ethyl sarcosinate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NHCH2COOC2H5 · HCl
CAS Number:
Molecular Weight:
153.61
Beilstein/REAXYS Number:
3911182
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Product Name

Sarcosine ethyl ester hydrochloride, puriss., ≥99.0% (AT)

grade

puriss.

assay

≥99.0% (AT)

form

crystals

ign. residue

≤0.05%

mp

~125 °C
127-128 °C (lit.)

solubility

H2O: 0.1 g/mL, clear

application(s)

peptide synthesis

SMILES string

Cl[H].CCOC(=O)CNC

InChI

1S/C5H11NO2.ClH/c1-3-8-5(7)4-6-2;/h6H,3-4H2,1-2H3;1H

InChI key

NIDZUMSLERGAON-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service