84509
SAOB solution
glycine buffer
Synonym(s):
Sulfide antioxidant solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
26111700
NACRES:
NB.61
form
solution
pH
13.5±0.3 ( in neat)
density
1.02 g/mL at 20 °C
General description
Visit our Sensor Applications portal to learn more.
Other Notes
This buffer prevents the oxidation of sulfide in the selective assay of sulfide ions with ISE
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
R. Bock et al.
Fresenius Journal of Analytical Chemistry, 240, 381-381 (1968)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service