84211
Salicylic acid
tested according to Ph. Eur.
Synonym(s):
Acidum salicylicum, 2-Hydroxybenzoic acid
About This Item
agency
USP/NF
tested according to Ph. Eur.
Quality Level
vapor density
4.8 (vs air)
vapor pressure
1 mmHg ( 114 °C)
bp
211 °C (lit.)
mp
158-161 °C (lit.)
SMILES string
OC(=O)c1ccccc1O
InChI
1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)
InChI key
YGSDEFSMJLZEOE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
314.6 °F - closed cup
flash_point_c
157 °C - closed cup
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service