Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C13H18O7
CAS Number:
Molecular Weight:
286.28
EC Number:
205-331-6
UNSPSC Code:
12352201
MDL number:
Beilstein/REAXYS Number:
89593
assay
≥99.0% (TLC)
optical activity
[α]20/D −62.5±2°, c = 3% in H2O
mp
196-202 °C
SMILES string
OC[C@H]1O[C@@H](Oc2ccccc2CO)[C@H](O)[C@@H](O)[C@@H]1O
InChI
1S/C13H18O7/c14-5-7-3-1-2-4-8(7)19-13-12(18)11(17)10(16)9(6-15)20-13/h1-4,9-18H,5-6H2/t9-,10-,11+,12-,13-/m1/s1
InChI key
NGFMICBWJRZIBI-UJPOAAIJSA-N
Other Notes
Substrate for β-glucosidase
Still not finding the right product?
Explore all of our products under D-(−)-Salicin
signalword
Warning
hcodes
pcodes
Hazard Classifications
Skin Sens. 1
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Beta-Glycosidase of potato.
P BARUAH et al.
The Biochemical journal, 66(2), 321-323 (1957-06-01)
Brian S Luisi et al.
Journal of pharmaceutical sciences, 101(4), 1475-1485 (2012-01-04)
During milling, components are subjected to shear and tensile stresses, which can result in physical phase transformations. The purpose of the work described in this report is to understand the pathway by which two test compounds, d-salicin and γ-indomethacin, undergo
Jeannette Y Wick
The Consultant pharmacist : the journal of the American Society of Consultant Pharmacists, 27(5), 322-329 (2012-05-18)
Most pharmacists know that aspirin's origins lie with willow bark, but they may be unaware of its role in the development of the pharmaceutical industry. Evolving from salacin (the active ingredient in many plant remedies) to salicylic acid (an analgesic
