Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

83926

Sigma-Aldrich

Rose oil, bulgarian from Rosa damascena Mill.

natural

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12164502
organoleptic:
rose

form

oil

quality

natural

refractive index

n20/D 1.46

application(s)

flavors and fragrances

organoleptic

rose

storage temp.

2-8°C

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

159.8 °F - closed cup

flash_point_c

71 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service