form
beads
technique(s)
LPLC: suitable
matrix active group
polymer
separation technique
size exclusion (SEC)
storage temp.
2-8°C
Features and Benefits
PD is a novel polymer offering properties similar to PS, but has a much lower oxygen permeability which results in a higher photostability for most dyes.
Physical form
0.5% solids in aqueous buffer, pH 7.0
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service