83130
Pyrogallol
purum, ≥98.0% (HPLC)
Synonym(s):
1,2,3-Trihydroxybenzene, 2,3-Dihydroxyphenol, Pyrogallic acid
About This Item
vapor density
4.4 (vs air)
vapor pressure
10 mmHg ( 167.7 °C)
grade
purum
assay
≥98.0% (HPLC)
ign. residue
≤0.05%
bp
309 °C (lit.)
mp
132-134 °C
133-134 °C (lit.)
SMILES string
Oc1cccc(O)c1O
InChI
1S/C6H6O3/c7-4-2-1-3-5(8)6(4)9/h1-3,7-9H
InChI key
WQGWDDDVZFFDIG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Muta. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service