Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

82611

Sigma-Aldrich

Pyrazinecarboxylic acid

purum, ≥98.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N2O2
CAS Number:
Molecular Weight:
124.10
Beilstein/REAXYS Number:
112305
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥98.0% (T)

ign. residue

≤0.1%

mp

222-225 °C (dec.) (lit.)

SMILES string

OC(=O)c1cnccn1

InChI

1S/C5H4N2O2/c8-5(9)4-3-6-1-2-7-4/h1-3H,(H,8,9)

InChI key

NIPZZXUFJPQHNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Iwanaga et al.
The Journal of pharmacology and experimental therapeutics, 320(1), 211-217 (2006-10-18)
Serum uric acid (SUA) is currently recognized as a risk factor for cardiovascular disease. It has been reported that an angiotensin II receptor blocker (ARB), losartan, decreases SUA level, whereas other ARBs, such as candesartan, have no lowering effect. Because
H J Kim et al.
The international journal of tuberculosis and lung disease : the official journal of the International Union against Tuberculosis and Lung Disease, 16(1), 98-103 (2012-01-13)
Pyrazinamide (PZA), one of the most effective anti-tuberculosis drugs, becomes toxic to Mycobacterium tuberculosis when converted to pyrazinoic acid by pyrazinamidase (PZase). PZA resistance is caused mainly by the loss of enzyme activity by mutation. To investigate the patterns of
Jhy-Wen Wu et al.
Drug metabolism and disposition: the biological fate of chemicals, 35(9), 1603-1610 (2007-06-02)
Pyrazinamide (PZA) is widely used in combination with other drugs in chemotherapy for tuberculosis. However, the dose-related liver injury is the main adverse effect of PZA and its metabolite [pyrazinoic acid (PA)]. Silibinin is the main flavonoid extracted from milk
Y Sheeena Mary et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(2), 725-730 (2008-03-21)
A substituted amide of pyrazine-2-carboxylic acid was prepared and the IR spectrum is recorded and analysed. The vibrational frequencies and corresponding vibrational assignments are examined theoretically using the Gaussian03 set of quantum chemistry codes. Predicted infrared and Raman intensities are
João P S Fernandes et al.
Archiv der Pharmazie, 343(2), 91-97 (2010-01-26)
Tuberculosis is an infection caused mainly by Mycobacterium tuberculosis. A first-line antimycobacterial drug is pyrazinamide (PZA), which acts partially as a prodrug activated by a pyrazinamidase releasing the active agent, pyrazinoic acid (POA). As pyrazinoic acid presents some difficulty to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service