Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

82492

Supelco

Protein A, immobilized on Agarose CL-4B from Staphylococcus aureus

suitable for affinity chromatography

Synonym(s):

Protein A-Agarose CL-4B

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12190000
NACRES:
NA.21

form

powder

Quality Level

contains

lactose as stabilizer

technique(s)

affinity chromatography: suitable

capacity

~20 mg/mL, gel binding capacity (human IgG)

Storage temp.

−20°C

General description

protein A is immobilized on cross-linked, CNBr-activated agarose 4B, which must be removed prior to use by washing gel onto filter; 1 g powder yields ~4 mlswollen gel

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service