Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

82487

Supelco

Protein A, immobilized on polyacrylic beads, from Staphylococcus aureus

suitable for affinity chromatography

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000

form

beads

technique(s)

LPLC: suitable
affinity chromatography: suitable

matrix

Polyacrylamide

matrix active group

Protein A

particle size

150 μm

capacity

~5 mg/mL, gel binding capacity (human IgG)

separation technique

affinity

storage temp.

−20°C

General description

protein A is immobilized on hydrophilic macroporous oxirane acrylic beads (particle size 150 μm) with an electroneutral mechanism for binding proteins via a C2-spacer

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service