Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

82486

Supelco

Protein A, immobilized on Agarose CL-4B, from Staphylococcus aureus

suitable for affinity chromatography

Synonym(s):

Protein A-Agarose CL-4B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000

form

suspension

contains

0.02% thimerosal as preservative

technique(s)

LPLC: suitable
affinity chromatography: suitable

capacity

~10 mg/mL, gel binding capacity (human IgG)

storage temp.

2-8°C

General description

protein A is immobilized on cross-linked 4-nitrophenylcarbonate-activated agarose 4B

Physical form

50% aqueous suspension in 0.9% sodium chloride

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wilchek, M. and Miron, T.
Biochemistry International, 4, 629-629 (1982)
Jelena Janković et al.
Disease markers, 33(6), 321-331 (2012-10-02)
We evaluated caveolin-1 expression in the human thyroid neoplasia spectrum with the aim of examining differences in expression as detected by two anti-caveolin-1 antibodies, and secondly, to investigate the association of caveolin-1 expression levels with aggressive papillary thyroid carcinoma (PTC).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service