Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

82325

Sigma-Aldrich

(±)-Propylene oxide

purum, ≥99.0% (GC)

Synonym(s):

(±)-Methyloxirane, 1,2-Epoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O
CAS Number:
Molecular Weight:
58.08
Beilstein/REAXYS Number:
79763
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor density

2 (vs air)

vapor pressure

29.43 psi ( 55 °C)
8.59 psi ( 20 °C)

grade

purum

assay

≥99.0% (GC)

form

liquid

autoignition temp.

1378 °F

expl. lim.

37 %

refractive index

n20/D 1.366 (lit.)
n20/D 1.366

bp

34 °C (lit.)

mp

−112 °C (lit.)

density

0.83 g/mL at 25 °C (lit.)

SMILES string

CC1CO1

InChI

1S/C3H6O/c1-3-2-4-3/h3H,2H2,1H3

InChI key

GOOHAUXETOMSMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 1 - Muta. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-36.4 °F - Equilibrium method

flash_point_c

-38 °C - Equilibrium method

ppe

Faceshields, Gloves, Goggles


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan Farjon et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 210(1), 24-30 (2011-03-08)
The use of chiral liquid crystals as NMR solvent is a powerful tool to visualize and quantify enantiomers. This technique is much more robust than traditional liquid state techniques. Recent developments have been performed to simplify proton spectra obtained in
Sascha Brunke et al.
PLoS pathogens, 10(10), e1004478-e1004478 (2014-10-31)
Candida glabrata is one of the most common causes of candidemia, a life-threatening, systemic fungal infection, and is surpassed in frequency only by Candida albicans. Major factors contributing to the success of this opportunistic pathogen include its ability to readily
Denis Merlet et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 209(2), 315-322 (2011-02-25)
The application of the G-SERFph pulse sequence is presented on enantiomeric mixtures dissolved in a chiral liquid crystal. It aims at editing, within one single 2D spectrum, every proton coupling which is experienced by a given proton site in the
Aude Bouchet et al.
The Journal of organic chemistry, 76(10), 4178-4181 (2011-04-08)
ECD and NMR experiments show that the complexation of propylene oxide (PrO) within the cavity of an enantiopure water-soluble cryptophane 1 in NaOH solution is enantioselective and that the (R)-PrO@PP-1 diastereomer is more stable than the (S)-PrO@PP-1 diastereomer with a
Yasuaki Tokudome et al.
Journal of colloid and interface science, 352(2), 303-308 (2010-09-09)
The structure formation process of hierarchically porous alumina gels has been investigated by in situ small angle X-ray scattering (SAXS). The measurement was performed on the sol-gel solution containing aluminum chloride hexahydrate (AlCl(3)·6H(2)O), poly(ethylene oxide) (PEO), and propylene oxide (PO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service