Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

81523

Supelco

Poly(α-methylstyrene)

analytical standard, 300,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2C(CH3)(C6H5)]n
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

mol wt

Mn ~427000
Mp ~440000
Mw ~437000

technique(s)

gel permeation chromatography (GPC): suitable

density

1.075 g/mL at 25 °C (lit.)

Mw/Mn

~1.02

format

neat

storage temp.

2-8°C

SMILES string

CC(=C)c1ccccc1

InChI

1S/C9H10/c1-8(2)9-6-4-3-5-7-9/h3-7H,1H2,2H3

InChI key

XYLMUPLGERFSHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service