Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

81421

Supelco

Polystyrene standard set Mp 400-1′800′000

suitable for gel permeation chromatography (GPC)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

technique(s)

gel permeation chromatography (GPC): suitable

format

neat

Components

set contains 12 different polystyrene standards with 1 g with Mp ~472 (Cat. No. 81401), ~1920 (Cat. No. 81403), ~3250 (Cat. No. 81404), ~10250 (Cat. No. 81406), ~24000 (Cat. No. 81407),~32500 (Cat. No. 81408), ~67500 (Cat. No. 81409), ~160000 (Cat. No. 81411), ~295000 (Cat. No. 81413), ~705000 (Cat. No. 81415), ~1000000 (Cat. No. 81416), ~1800000 (Cat. No. 92847); The components can also be ordered separately under the product numbers indicated.

Storage Class

10 - Combustible liquids

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service