Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

81303

Supelco

Polyaniline (emeraldine salt)

Selectophore

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
26111700
NACRES:
NB.61

product line

Selectophore

Quality Level

form

powder

mp

>300 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

polymeric-membrane material for chemo- and biosensoric applications[1]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A. Aytac et al.
Russ. J. Electrochem., 40, 732-732 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service