Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

81219

Supelco

Polyethylene

analytical standard, suitable for gel permeation chromatography (GPC), 1,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(CH2CH2)nH
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

mol wt

Mn ~1110
Mp ~1180
Mw ~1230

technique(s)

gel permeation chromatography (GPC): suitable

Mw/Mn

~1.11

format

neat

storage temp.

2-8°C

InChI

1S/C2H4/c1-2/h1-2H2

InChI key

VGGSQFUCUMXWEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahtab Memarpour et al.
The journal of adhesive dentistry, 15(1), 85-91 (2013-03-28)
To evaluate the clinical performance of polyethylene fiber for anterior primary teeth restorations in children after 2.5 years, according to modified Ryge criteria. Fifty-five maxillary incisors with extensive caries in 24 Iranian patients (mean age 4.2 years) were restored. All
Lisa K McCarthy et al.
Pediatrics, 132(1), e135-e141 (2013-06-19)
Hypothermia on admission to the NICU is associated with increased mortality in preterm infants. Many newborns are hypothermic on admission despite using polyethylene bags (PBs). Using exothermic mattresses (EMs) in addition to PBs may reduce hypothermia but increase hyperthermia. We
[Technologies for manufacturing ultra-high molecular polyethylene-based porous structures for bone implants].
A V Maksimkin et al.
Meditsinskaia tekhnika, (2)(2), 16-20 (2013-06-26)
Frederick J Kummer et al.
Journal of long-term effects of medical implants, 22(2), 177-179 (2013-02-23)
A polyethylene cup explanted after 41 years was examined using several analytical techniques to determine whether there was a material cause for the extremely low wear observed. Neither the amount of polyethylene oxidation nor crystallinity appeared to be a factor.
Matteo Gumiero et al.
Food chemistry, 138(2-3), 1633-1640 (2013-02-16)
A high density polyethylene (HDPE)/calcium carbonate (CaCO(3)) film containing TiO(2) was prepared via blown film extrusion process. The photocatalytic properties of this film were evaluated by voltammetric, UV-Vis spectrophotometric and gas chromatographic measurements following the decomposition rate of suitably selected

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service