Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

81162

Sigma-Aldrich

Poly(ethylene glycol)

tested according to Ph. Eur., 300

Synonym(s):

Macrogol 300, Polyoxyethylene, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21

agency

tested according to Ph. Eur.

Quality Level

form

liquid

mol wt

Mr 285-315

pH

6-7 (23 °C, at 10 g/l)

application(s)

pharmaceutical (small molecule)

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) is a linear polyether of ethylene glycol (ethane-1,2-diol).[1] This non-ionic polymer[2] is soluble in water, benzene, ethanol, acetonitrile, and dichloromethane. It is insoluble in diethyl ether and hexane.[3] It usually exists in the monomer and aqueous phases.[4]

Application

Poly(ethylene glycol) has been used:
  • to dissolve NEO2734 (BET and CBP/EP300 inhibitor) for drug efficacy studies in vivo[5]
  • as a drug carrier for histone deacetylase (HDAC) inhibitor ACY-1215[6]
  • to study the effects of drug-induced peripheral neuropathy[7]

Biochem/physiol Actions

Poly(ethylene glycol) (PEG) is used in various pharmaceutical applications.[3] This polymer occurs as a preservative and is used in biomedical applications like drug delivery systems, biosensing, and tissue engineering. PEG is also used in cosmetic ointments and creams. It is used as a binding and dispersing agent in chemical and other industries.[8]

Features and Benefits

  • Environment friendly[4]
  • Biocompatible[4]
  • Unique solvent solution[4]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

428.0 °F - closed cup

flash_point_c

220.00 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymers for 3D bioprinting
Beata K
Journal of Separation Science, 337-349 (2022)
Timothy A Yap et al.
Nature medicine, 29(1), 115-126 (2023-01-20)
Although targeting oxidative phosphorylation (OXPHOS) is a rational anticancer strategy, clinical benefit with OXPHOS inhibitors has yet to be achieved. Here we advanced IACS-010759, a highly potent and selective small-molecule complex I inhibitor, into two dose-escalation phase I trials in patients with
Application of compatibilized polymer blends in biomedical fields
Zarrintaj P, et al.
Journal of Separation Science, 511-537 (2020)
Polymers and polymeric hybrids for targeted drug delivery
Islam R and Fang J
Journal of Separation Science, 303-322 (2022)
Yiming Shi et al.
Langmuir : the ACS journal of surfaces and colloids, 29(9), 3024-3033 (2013-02-26)
Through zeta potential and surface tension measurements and a series of polymerization experiments, the role of poly(ethylene glycol) (PEG) in the process of surfactant-free polymerization of styrene (St)/methyl methacrylate (MMA) has been investigated experimentally. Nanoscale and stable copolymer particles were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service