Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80779

Supelco

Gymnodimine solution

20 μg/g in methanol (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H45NO4
CAS Number:
Molecular Weight:
507.70
MDL number:
UNSPSC Code:
41116107

grade

certified reference material
TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

20 μg/g in methanol (nominal concentration)

application(s)

cleaning products
cosmetics
food and beverages
personal care

storage temp.

−20°C

InChI

1S/C32H45NO4/c1-18-14-27-22(5)26(30-16-21(4)32(35)37-30)10-8-23(27)17-24-7-6-12-33-28(24)15-20(3)31-19(2)13-25(36-31)9-11-29(18)34/h14-16,19,23-25,27,29-31,34H,6-13,17H2,1-5H3/b18-14+,20-15+/t19-,23?,24-,25+,27+,29+,30+,31+/m1/s1

InChI key

UCHGCIXABSCOFK-STHGLBKMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI.
Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

For the analysis of marine toxins in seafood.

Check out our complete range of marine toxins reference materials

Analysis Note

20 μg/g in 35 μmol/l AcOH in methanol

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service