Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80774

Supelco

O-Demethylmetoprolol

analytical standard

Synonym(s):

3-[4-(2-Hydroxyethyl)phenoxy]-1-(N-isopropyl)aminopropan-2-ol, 4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzeneethanol, O-Desmethylmetoprolol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H23NO3
CAS Number:
Molecular Weight:
253.34
Beilstein/REAXYS Number:
8416818
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥97% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical

format

neat

SMILES string

OCCC1=CC=C(OCC(CNC(C)C)O)C=C1

InChI

1S/C14H23NO3/c1-11(2)15-9-13(17)10-18-14-5-3-12(4-6-14)7-8-16/h3-6,11,13,15-17H,7-10H2,1-2H3

InChI key

CUKXSBOAIJILRY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazardEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Repr. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alfred Svan et al.
Journal of mass spectrometry : JMS, 51(3), 207-218 (2016-03-10)
Identification of degradation products from trace organic compounds, which may retain the biological activity of the parent compound, is an important step in understanding the long-term effects of these compounds on the environment. Constructed wetlands have been successfully utilized to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service