Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80693

Supelco

(±)-2-Propyl-4-pentenoic acid

analytical standard

Synonym(s):

2-Allylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O2
CAS Number:
Molecular Weight:
142.20
Beilstein/REAXYS Number:
1704069
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

C=CCC(CCC)C(O)=O

InChI

1S/C8H14O2/c1-3-5-7(6-4-2)8(9)10/h3,7H,1,4-6H2,2H3,(H,9,10)

InChI key

UMYDNZXEHYSVFY-UHFFFAOYSA-N

General description

2-Propyl-4-pentenoic acid is the main teratogenic and hepatotoxic metabolite of valproic acid, an anticonvulsant prescribed for treating seizures and for the maintenance phase of bipolar disorders.[1][2][3]

Application

2-Propyl-4-pentenoic acid may be used as an analytical standard for the determination of the analyte in biological samples[4][5] by various chromatography-based techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of valproic acid and its metabolite 2-propyl-4-pentenoic acid in human plasma using HPLC-MS/MS
Cheng H, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 850(1-2), 206-212 (2007)
Determination of valproic acid and its metabolites using gas chromatography with mass-selective detection: application to serum and urine samples from sheep
Yu D, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 666(2), 269-281 (1995)
Comparative toxicity of valproic acid and its metabolites in liver slices from adult rats, weanling rats and humans
Fisher RL, et al.
Toxicology in vitro, 8(3), 371-379 (1994)
Cheng Chang et al.
Molecular pharmacology, 94(2), 823-833 (2018-06-02)
Kynurenic acid (KYNA) plays a significant role in maintaining normal brain function, and abnormalities in KYNA levels have been associated with various central nervous system disorders. Confirmation of its causality in human diseases requires safe and effective modulation of central
Prophylactic efficacy of lithium, valproic acid, and carbamazepine in the maintenance phase of bipolar disorder: a naturalistic study
Peselow ED, et al.
International Clinical Psychopharmacology, 31(4), 218-223 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service