Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80642

Sigma-Aldrich

Piperidine

purum, ≥96.0% (GC/T)

Synonym(s):

Hexahydropyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11N
CAS Number:
Molecular Weight:
85.15
Beilstein/REAXYS Number:
102438
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3 (vs air)

vapor pressure

23 mmHg ( 20 °C)

grade

purum

assay

≥96.0% (GC/T)

form

liquid

impurities

≤0.5% water

refractive index

n20/D 1.452 (lit.)
n20/D 1.453

bp

106 °C (lit.)

mp

−13 °C (lit.)

solubility

organic solvents: soluble(lit.)
water: miscible(lit.)

density

0.862 g/mL at 20 °C (lit.)

SMILES string

C1CCNCC1

InChI

1S/C5H11N/c1-2-4-6-5-3-1/h6H,1-5H2

Inchi Key

NQRYJNQNLNOLGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

60.8 °F - closed cup

flash_point_c

16 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moessbauer spectra of some porphyrin complexes with pyridine, piperidine, and imidazole.
Epstein LM, et al.
Inorganic Chemistry, 6(9), 1720-1724 (1967)
Simultaneous recrystallization and post-synthesis of MCM-49 zeolite by TEAOH/piperidine: Implication on the acidity and catalytic alkylation performance.
Shi Y, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 220, 7-15 (2016)
Marek Łuczkowski et al.
Inorganic chemistry, 47(23), 10875-10888 (2008-10-31)
A de novo protein design strategy provides a powerful tool to elucidate how heavy metals interact with proteins.Cysteine derivatives of the TRI peptide family (Ac-G(LKALEEK)4G-NH2) have been shown to bind heavy metals in an unusual trigonal geometry. Our present objective
Total synthesis of (-)-kopsinine by an asymmetric one-pot [n+2+3] cyclization.
Shingo Harada et al.
Chemistry, an Asian journal, 7(10), 2196-2198 (2012-08-22)
J Phillip Turner et al.
ACS chemical neuroscience, 5(7), 552-558 (2014-04-03)
Alzheimer's disease (AD) is the most common form of dementia and the sixth leading cause of death in the United States. Plaques composed of aggregated amyloid-beta protein (Aβ) accumulate between the neural cells in the brain and are associated with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service