Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80595

Supelco

(1S)-(−)-trans-Pinane

analytical standard

Synonym(s):

(1S,2S)-2,6,6-Trimethylbicyclo[3.1.1]heptane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18
CAS Number:
Molecular Weight:
138.25
Beilstein/REAXYS Number:
2321791
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.5% (sum of enantiomers, GC)

optical activity

[α]20/D −17±1°, neat

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.461

bp

165-167 °C (lit.)

density

0.854 g/mL at 20 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

[H][C@]12CC[C@H](C)[C@]([H])(C1)C2(C)C

InChI

1S/C10H18/c1-7-4-5-8-6-9(7)10(8,2)3/h7-9H,4-6H2,1-3H3/t7-,8-,9-/m0/s1

InChI key

XOKSLPVRUOBDEW-CIUDSAMLSA-N

application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

118.4 °F - closed cup

flash_point_c

48 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service