Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

80112

Supelco

Serdolit CG-400 II

Cl--form, strongly basic, 200-400 mesh

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.52

form

beads

technique(s)

LPLC: suitable

loss

≤20% loss on drying, 110°C

matrix

styrene-divinylbenzene

matrix active group

quaternary ammonium

particle size

200-400 mesh

capacity

1.9 meq/mL

separation technique

anion exchange

Legal Information

Serdolit is a trademark of Serva Electrophoresis GmbH

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service