Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

79890

Millipore

Semi-solid nutrient agar ISO 6579:2002

NutriSelect® Plus, suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

agar, 4.0 g/L
beef extract, 3.0 g/L
peptic digest of animal tissue, 5.0 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.0±2.0 (25 °C)

application(s)

environmental
food and beverages

microbiology

suitability

Salmonella spp.

Application

Semisolid Nutrient Agar is recommended for detection of Salmonella species on the basis of motility and hydrogen sulphide (H2S) production.

Preparation Note

Suspend 12 grams in 1000 ml distilled water. Heat to boiling to dissolve the medium completely. Dispense in tubes. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. Allow the tubes to cool in an upright position.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service