Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

79743

Supelco

Carbon tetrachloride

suitable for IR spectroscopy, ≥99.9% (GC)

Synonym(s):

Tetrachloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CCl4
CAS Number:
Molecular Weight:
153.82
Beilstein/REAXYS Number:
1098295
EC Number:
MDL number:
UNSPSC Code:
41116004

vapor density

5.32 (vs air)

Quality Level

vapor pressure

143 mmHg ( 30 °C)
91 mmHg ( 20 °C)

assay

≥99.9% (GC)

technique(s)

IR spectroscopy: suitable

impurities

≤0.00005% free chlorine (as Cl)
≤0.0002% free acid (as HCl)
≤0.005% water
≤1 ppb fluorescence (quinine) at 254 nm
≤1 ppb fluorescence (quinine) at 365 nm

evapn. residue

≤0.0005%

refractive index

n20/D 1.459-1.461
n20/D 1.460 (lit.)

bp

76-77 °C (lit.)

mp

−23 °C (lit.)

density

1.594 g/mL at 25 °C (lit.)

UV absorption

λ: 265 nm Amax: ≤0.698
λ: 270 nm Amax: ≤0.301
λ: 280 nm Amax: ≤0.045
λ: 290 nm Amax: ≤0.008
λ: 295 nm Amax: ≤0.004

SMILES string

ClC(Cl)(Cl)Cl

InChI

1S/CCl4/c2-1(3,4)5

InChI key

VZGDMQKNWNREIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Ozone 1 - Skin Sens. 1B - STOT RE 1 Inhalation

target_organs

Liver,Kidney

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service