Skip to Content
MilliporeSigma

79510

Sodium phosphonoformate tribasic hexahydrate

≥97.0% (NT)

Synonym(s):

Foscarnet, Phosphonoformic acid trisodium salt hexahydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Na2O3PCO2Na · 6H2O
CAS Number:
Molecular Weight:
300.04
UNSPSC Code:
12352100
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥97.0% (NT)

impurities

35-37% water

solubility

H2O: 0.1 g/mL, clear, colorless (hot)

SMILES string

O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]C(=O)P([O-])([O-])=O

InChI

1S/CH3O5P.3Na.6H2O/c2-1(3)7(4,5)6;;;;;;;;;/h(H,2,3)(H2,4,5,6);;;;6*1H2/q;3*+1;;;;;;/p-3

InChI key

ILRVASBWNRYBFD-UHFFFAOYSA-K

Gene Information

Other Notes

Antiviral agent


pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Muta. 2 - STOT RE 2

target_organs

Kidney,Bone

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library