Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

79449

Supelco

Diatoxanthin

analytical standard

Synonym(s):

(3R,3′R)-7,8-Didehydro-β,β-carotene-3,3′-diol, all-trans-Diatoxanthin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H54O2
CAS Number:
Molecular Weight:
566.86
Beilstein/REAXYS Number:
1917792
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

Quality Level

assay

≥94.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

−20°C

InChI

1S/C40H54O2/c1-29(17-13-19-31(3)21-23-37-33(5)25-35(41)27-39(37,7)8)15-11-12-16-30(2)18-14-20-32(4)22-24-38-34(6)26-36(42)28-40(38,9)10/h11-21,23,35-36,41-42H,25-28H2,1-10H3/b12-11+,17-13+,18-14+,23-21+,29-15+,30-16+,31-19+,32-20+/t35-,36-/m1/s1

InChI key

HNYJHQMUSVNWPV-DRCJTWAYSA-N

General description

Diatoxanthin belongs to the class of carotenoids.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Gottardis et al.
Der Anaesthesist, 36(3), 132-136 (1987-03-01)
Acute changes involving the hormones thyroxine (T4) and triiodothyronine (T3) followed by hyper -or hypothyroid crises will lead to both cardiovascular and metabolic disorders. Due to highly sensitive mechanisms controlling central hormonal production and peripheral enzymatic conversion (c.g. T4 to
L Van Heukelem et al.
Journal of chromatography. A, 910(1), 31-49 (2001-03-27)
We used chromatography modeling software to assist in HPLC method development, with the goal of enhancing separations through the exclusive use of gradient time and column temperature. We surveyed nine stationary phases for their utility in pigment purification and natural
M C Cerón-García et al.
Food chemistry, 257, 316-324 (2018-04-07)
Microalgae are an interesting source of natural pigments that have valuable applications. However, further research is necessary to develop processes that allow us to achieve high levels of carotenoid recovery while avoiding degradation. This work presents a comprehensive study on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service