Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

79379

Sigma-Aldrich

Phosphate citrate buffer with hydrogen peroxide urea adduct

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700

solubility

H2O: 1 tablet/100mL, clear, colorless

storage temp.

2-8°C

Application

application procedure: dissolve 1 tablet in 100 mL deionized water in order to get a 0.05 M buffer solution containing 0.014% H2O2, pH 5.0 at 25°C

Packaging

One package contains 50 tablets.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service