Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

79180BC

Millipore

GDS PPMX Tip Box Holder

BioControl Systems, for use with GDS PickPen® PIPETMAX® System

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$654.00

$654.00


Available to ship onApril 29, 2025Details


Request more information

Select a Size

Change View
1 EACH
$654.00

About This Item

UNSPSC Code:
41121521
NACRES:
NA.85

$654.00


Available to ship onApril 29, 2025Details


Request more information

manufacturer/tradename

BioControl Systems

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS PickPen PIPETMAX System

storage temp.

room temp

Legal Information

PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PIPETMAX is a registered trademark of Gilson, Inc

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service