Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78833

Supelco

2-Methylthiophene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6S
CAS Number:
Molecular Weight:
98.17
Beilstein/REAXYS Number:
103734
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Quality Level

vapor pressure

47.6 mmHg ( 37.7 °C)

assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.519-1.521
n20/D 1.52 (lit.)

bp

113 °C (lit.)

mp

−63 °C (lit.)

density

1.014 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

Cc1cccs1

InChI

1S/C5H6S/c1-5-3-2-4-6-5/h2-4H,1H3

InChI key

XQQBUAPQHNYYRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Flame

signalword

Danger

hcodes

pcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

59.0 °F - closed cup

flash_point_c

15 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stéphanie Conotte et al.
Respiratory physiology & neurobiology, 256, 157-173 (2018-03-10)
Chronic intermittent hypoxia (ChIH) is a dominant feature of obstructive sleep apnoea (OSA) and is associated to metabolic alterations and oxidative stress (OS). Although management of OSA is well established, the research of new biomarkers that are independent of confounding
Boxing Li et al.
Cell, 181(7), 1547-1565 (2020-06-04)
Homeostasis of neural firing properties is important in stabilizing neuronal circuitry, but how such plasticity might depend on alternative splicing is not known. Here we report that chronic inactivity homeostatically increases action potential duration by changing alternative splicing of BK
Jennifer Maning et al.
International journal of molecular sciences, 21(8) (2020-04-25)
Aldosterone (Aldo), when overproduced, is a cardiotoxic hormone underlying heart failure and hypertension. Aldo exerts damaging effects via the mineralocorticoid receptor (MR) but also activates the antiapoptotic G protein-coupled estrogen receptor (GPER) in the heart. G protein-coupled receptor (GPCR)-kinase (GRK)-2
Xiaodong Feng et al.
Cancer cell, 35(3), 457-472 (2019-02-19)
Activating mutations in GNAQ/GNA11, encoding Gαq G proteins, are initiating oncogenic events in uveal melanoma (UM). However, there are no effective therapies for UM. Using an integrated bioinformatics pipeline, we found that PTK2, encoding focal adhesion kinase (FAK), represents a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service