Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78700

Supelco

Methyl 2-octynoate

reference material

Synonym(s):

Methyl heptine carbonate, NSC 72098

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4C≡CCO2CH3
CAS Number:
Molecular Weight:
154.21
Beilstein/REAXYS Number:
1756887
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

reference material

Quality Level

assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.446 (lit.)

bp

217-220 °C (lit.)

density

0.92 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCCC#CC(=O)OC

InChI

1S/C9H14O2/c1-3-4-5-6-7-8-9(10)11-2/h3-6H2,1-2H3

InChI key

FRLZQXRXIKQFNS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.

Methyl 2-octynoate, belonging to the class of acetylenic esters,[1] is an allergen found in cosmetics,[2] often used for green, violet-leaf, floral notes in fragrances.[1]

Application

Methyl 2-octynoate may be used as a fragrance allergen standard for the quantification of the analyte in cosmetics[3][2][4] using chromatography techniques.

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Skin Sens. 1A

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

192.2 °F - closed cup

flash_point_c

89 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of suspected fragrance allergens in cosmetics by matrix solid-phase dispersion gas chromatography--mass spectrometry analysis
Sanchez-Prado L, et al.
Journal of Chromatography A, 1218(31), 5055-5062 (2011)
Aroma Chemicals I: C, H, O Compounds
Chemistry and Technology of Flavors and Fragrances., 72-73 (2005)
Miguel del Nogal Sánchez et al.
Analytical and bioanalytical chemistry, 397(6), 2579-2591 (2010-05-25)
In the present work, a strategy for the qualitative and quantitative analysis of 24 volatile compounds listed as suspected allergens in cosmetics by the European Union is reported. The list includes benzyl alcohol, limonene, linalool, methyl 2-octynoate, beta-citronellol, geraniol, citral
Validation of SPME-GC-FID method for determination of fragrance allergens in selected cosmetic products
Divisova R, et al.
Acta Chromatographica , 27(3), 509-523 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service