Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78690

Supelco

Phenylhydrazine hydrochloride

for the detection of sugar, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHNH2 · HCl
CAS Number:
Molecular Weight:
144.60
Beilstein/REAXYS Number:
3594958
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
NACRES:
NA.21

Quality Level

assay

≥99.0% (AT)
≥99.0%

form

powder

quality

for the detection of sugar

ign. residue

≤0.05%

mp

250-254 °C (dec.) (lit.)

application(s)

food and beverages
general analytical

SMILES string

Cl[H].NNc1ccccc1

InChI

1S/C6H8N2.ClH/c7-8-6-4-2-1-3-5-6;/h1-5,8H,7H2;1H

InChI key

JOVOSQBPPZZESK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M D Powell et al.
Journal of fish biology, 81(1), 335-344 (2012-07-04)
Phenylhydrazine injections (0.3 mg kg(-1) , followed by a second injection of 0.1 mg kg(-1) 7 days later) induced a reproducible and stable anaemia in Atlantic halibut Hippoglossus hippoglossus, reducing the haematocrit and haemoglobin by 70.0 and 75.5%, respectively, over
Ahmed A Ibrahim et al.
Talanta, 93, 257-263 (2012-04-10)
We report here the fabrication of a robust, highly sensitive, reliable and reproducible phenyl hydrazine chemical sensor using Ag-doped ZnO nanoflowers as efficient electron mediators. The Ag-doped ZnO nanoflowers were synthesized by facile hydrothermal process at low-temperature and characterized in
Denham G Cook et al.
The Journal of experimental biology, 214(Pt 17), 2927-2934 (2011-08-13)
The effect of altered oxygen transport potential on behavioural responses to environmental hypoxia was tested experimentally in snapper, Pagrus auratus, treated with a haemolytic agent (phenylhydrazine) or a sham protocol. Standard metabolic rate was not different between anaemic and normocythaemic
Robert Moreau et al.
PloS one, 7(9), e46101-e46101 (2012-10-03)
Sustained erythropoiesis and concurrent bone marrow hyperplasia are proposed to be responsible for low bone mass density (BMD) in chronic hemolytic pathologies. As impaired erythropoiesis is also frequent in these conditions, we hypothesized that free heme may alter marrow and
Yuji Sonoda et al.
Cells, tissues, organs, 196(6), 555-564 (2012-06-29)
Fetal liver hematopoiesis in mice disappears approximately 2 weeks after birth; however, under experimental acute anemia extramedullary hematopoiesis occurs in the livers of adult mice. The hematopoietic foci in the liver during extramedullary hematopoiesis contain erythroblasts and macrophages. In this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service