Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78612

Sigma-Aldrich

Hexamethyldisilane

Wacker Chemie AG, ≥98.0% (GC)

Synonym(s):

Disilane M6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(Si(CH3)3)2
CAS Number:
Molecular Weight:
146.38
Beilstein/REAXYS Number:
1633463
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

≥98.0% (GC)

form

liquid

manufacturer/tradename

Wacker Chemie AG

refractive index

n20/D 1.422 (lit.)

bp

112-114 °C (lit.)

mp

9-12 °C (lit.)

density

0.715 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)[Si](C)(C)C

InChI

1S/C6H18Si2/c1-7(2,3)8(4,5)6/h1-6H3

InChI key

NEXSMEBSBIABKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

prices for bulk quantities on request

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

21.2 °F

flash_point_c

-6 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service