Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78262

Sigma-Aldrich

(R)-Oxynitrilase, Prunus amygdalus, CLEA

≥0.1 U/mg

Synonym(s):

(R)-Oxynitrilase, Prunus amygdalus, cross-linked enzyme aggregate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

powder

specific activity

≥0.1 U/mg

storage temp.

2-8°C

Unit Definition

1 U corresponds to the amount of enzyme which hydrolyzes 1μmol DL-mandelonitrile per minute at pH 3.8 and 30 °C

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service