Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78040

Sigma-Aldrich

DL-Phenylalanine

puriss., ≥99.0% (NT)

Synonym(s):

(±)-2-Amino-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
165.19
Beilstein/REAXYS Number:
3198807
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

puriss.

assay

≥99.0% (NT)

form

powder

ign. residue

≤0.05%

loss

≤0.1% loss on drying

mp

266-267 °C (dec.) (lit.)
271-273 °C (dec.)

application(s)

peptide synthesis

SMILES string

NC(Cc1ccccc1)C(O)=O

InChI

1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)

InChI key

COLNVLDHVKWLRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dávid Virág et al.
Journal of pharmaceutical and biomedical analysis, 180, 113018-113018 (2019-12-19)
Altered serotonergic neurotransmission is a key factor in several neurologic and psychiatric disorders such as migraine. Human and animal studies suggest that chronically low interictal serotonin levels of plasma and brain may facilitate increased activity of the trigeminovascular pathway, and
Sohei Nakayama et al.
Cancer research, 74(20), 5891-5902 (2014-08-29)
The discovery of somatic mutations in EGFR and development of EGFR tyrosine kinase inhibitors (TKI) have revolutionized treatment for lung cancer. However, resistance to TKIs emerges in almost all patients and currently no effective treatment is available. Here, we show
I-Ping Lee et al.
PloS one, 9(10), e109803-e109803 (2014-10-10)
The obligate intracellular parasite, Toxoplasma gondii, disseminates through its host inside infected immune cells. We hypothesize that parasite nutrient requirements lead to manipulation of migratory properties of the immune cell. We demonstrate that 1) T. gondii relies on glutamine for
Anke C Fender et al.
Naunyn-Schmiedeberg's archives of pharmacology, 387(10), 1001-1007 (2014-08-02)
Activators of protease-activated receptors PAR-1 and PAR-2 such as thrombin and synthetic hexapeptides promote hypertrophy of isolated neonatal cardiomyocytes at pathological concentrations. Since PAR-activating proteases often show dual actions at low vs. high concentrations, the potential hypertrophic effects of low-level
Atsushi Kanda et al.
The British journal of nutrition, 111(12), 2067-2078 (2014-03-07)
We have previously shown that whey protein hydrolysate (WPH) causes a greater increase in muscle protein synthesis than does a mixture of amino acids that is identical in amino acid composition. The present study was conducted to investigate the effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service