Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

77849

Supelco

(S)-(−)-1-Phenylethanol

derivatization grade (chiral), LiChropur, ≥99.0%

Synonym(s):

(−)-Methyl phenyl carbinol, (S)-(−)-α-Methylbenzyl alcohol, (S)-(−)-sec-Phenylethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10O
CAS Number:
Molecular Weight:
122.16
Beilstein/REAXYS Number:
2039797
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

derivatization grade (chiral)

Quality Level

product line

ChiraSelect

assay

≥99.0% (sum of enantiomers, GC)
≥99.0%

form

liquid

optical activity

[α]20/D −45±1°, c = 5% in methanol

optical purity

enantiomeric ratio: ≥98.5:1.5 (GC)

quality

LiChropur

technique(s)

HPLC: suitable

refractive index

n20/D 1.528

bp

88-89 °C/10 mmHg (lit.)

mp

9-11 °C (lit.)

density

1.012 g/mL at 20 °C (lit.)

SMILES string

C[C@H](O)c1ccccc1

InChI

1S/C8H10O/c1-7(9)8-5-3-2-4-6-8/h2-7,9H,1H3/t7-/m0/s1

InChI key

WAPNOHKVXSQRPX-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(−)-1-Phenylethanol is a high quality, useful chiral derivatization reagent for all your analytical applications, specific to GC in the chiral field. It is specially selected to meet the requirements for derivatization reagents for enantiomeric excess determinations.
(S)-(-)-1-Phenylethanol also known as (-)-Methyl phenyl carbinol is formed from reduction of acetophenone.[1]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Studies in detoxication. 59. The metabolism of alkylbenzenes; the biological reduction of ketones derived from alkylbenzenes.
SMITH JN, SMITHIES RH, WILLIAMS RT.
The Biochemical Journal, 57(1), 74-76 (1954)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service