Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

77686

Supelco

Biacangelicin

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18O7
CAS Number:
Molecular Weight:
334.32
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% water

application(s)

food and beverages

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C1C=CC(C(OC)=C(C=CO2)C2=C3OC[C@H](C(O)(C)C)O)=C3O1

InChI

1S/C17H18O7/c1-17(2,20)11(18)8-23-16-14-10(6-7-22-14)13(21-3)9-4-5-12(19)24-15(9)16/h4-7,11,18,20H,8H2,1-3H3/t11-/m1/s1

InChI key

PKRPFNXROFUNDE-LLVKDONJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyuncheol Oh et al.
Planta medica, 68(5), 463-464 (2002-06-12)
Fractionation of the MeOH extract of Angelica dahurica Benth et Hook resulted in the isolation of six furocoumarins, imperatorin (1), isoimperatorin (2), (+/-)-byakangelicol (3), (+)-oxypeucedanin (4), (+)-byakangelicin (5), and (+)-aviprin (6). Among these, compounds 1 and 5 exhibited strong hepatoprotective
Yong-qing Xiao et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 29(7), 654-657 (2004-10-27)
To work out a qualitay standard of Radix Angelicae dahuricae. TLC and HPLC were adopted. Four coumerin compounds were identified by TLC, and the contents of two compounds were determined by HPLC. Imperatorin, isoimperatorin, byakangelicin and oxypeucedaninhydrate can be found
Jian Yang et al.
British journal of pharmacology, 162(2), 441-451 (2010-10-15)
Byakangelicin is found in extracts of the root of Angelica dahurica, used in Korea and China as a traditional medicine to treat colds, headache and toothache. As byakangelicin can inhibit the effects of sex hormones, it may increase the catabolism
Ying Xie et al.
Journal of pharmaceutical and biomedical analysis, 44(1), 166-172 (2007-03-09)
A high-performance liquid chromatographic method was developed and validated for the determination and pharmacokinetic study of oxypeucedanin hydrate and byak-angelicin after oral administration of Angelica dahurica extracts in mongrel dog plasma. The coumarin components and the internal standard isopsoralen were
Yang Bae Kim et al.
Archives of pharmacal research, 25(3), 275-279 (2002-07-24)
The crystal structure of byakangelicin, one of furanocoumarin aldose reductase inhibitors, was determined by X-ray diffraction method. The crystal is triclinic, with a = 8.114(1), b = 10.194(1), c = 11.428(1)A, a = 111.50(1), beta= 95.57(1), gamma = 112.52(1) degrees

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service