Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

77646

Supelco

α-Cyano-2,4-difluorocinnamic acid

≥95.0% (HPLC)

Synonym(s):

α-Cyano-o,p-difluorocinnamic acid, 2-Cyano-3-(2,4-difluorophenyl)-2-propenoic acid, DiFCCA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5F2NO2
CAS Number:
Molecular Weight:
209.15
Beilstein/REAXYS Number:
22870653
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.21

Quality Level

assay

≥95.0% (HPLC)

form

crystals

analyte functional class(es)

drugs of abuse, ionic liquids (quantification)

analyte chemical class(es)

chlorinated lipids, lipids, peptides, phospholipids, phosphopeptides

technique(s)

MALDI-MS: suitable
collision-induced dissociation MS/MS (CID-MS/MS): suitable
matrix-enhanced secondary ion MS (ME-SIMS): suitable

solubility

methanol: 100 mg/10 mL, clear, colorless to light yellow

suitability

suitable for matrix substance for MALDI-MS

SMILES string

OC(/C(C#N)=C/C1=CC=C(F)C=C1F)=O

InChI

1S/C10H5F2NO2/c11-8-2-1-6(9(12)4-8)3-7(5-13)10(14)15/h1-4H,(H,14,15)/b7-3+

InChI key

UQLFCBBYHQBVQY-XVNBXDOJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

suitable as MALDI-Matrix material

related product

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcel Wiegelmann et al.
Analytical and bioanalytical chemistry, 405(22), 6925-6932 (2012-10-16)
A high analytical sensitivity in ultraviolet matrix-assisted laser desorption ionization mass spectrometry (MALDI-MS) is only achieved if the laser wavelength corresponds to a high optical absorption of the matrix. Laser fluence and the physicochemical properties of the compounds, e.g., the
Thorsten Jaskolla et al.
Journal of the American Society for Mass Spectrometry, 20(5), 867-874 (2009-02-10)
Phosphatidylethanolamines (PEs) are abundant lipid constituents of the cellular membrane. The amino group of PEs exhibits high reactivity with hypochlorous acid that is generated under inflammatory conditions in vivo. The analysis of the resulting PE mono- and dichloramines is of
Dimitrios G Papasotiriou et al.
Journal of proteome research, 9(5), 2619-2629 (2010-04-07)
Peptide Mass Fingerprinting (PMF) of tryptically in-gel digested samples is a well-established protein identification technique for MALDI mass spectrometry but an in-depth PMF evaluation for in-gel digestions of less specific enzymes is still missing. This study demonstrates that the MALDI-LTQ-Orbitrap
Mark W Towers et al.
Journal of proteome research, 9(4), 1931-1940 (2010-02-13)
Matrix-assisted laser desorption/ionization (MALDI) is a key ionization technique in mass spectrometry (MS) for the analysis of labile macromolecules. An important area of study and improvements in relation to MALDI and its application in high-sensitivity MS is that of matrix
Thorsten W Jaskolla et al.
Journal of proteome research, 8(7), 3588-3597 (2009-05-14)
The performance of the recently developed 4-chloro-alpha-cyanocinnamic acid (Cl-CCA) matrix-assisted laser desorption ionization mass spectrometry (MALDI MS) matrix was investigated in comparison to the most widely used matrix alpha-cyano-4-hydroxycinnamic acid (CHCA). For this purpose, in-solution digestions of standard proteins in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service